Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium 1-Octadecanesulfonate, TCI America™
SDP

Catalog No. O01245G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O01245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O01245G Supplier TCI America Supplier No. O01245G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13893-34-0
Molecular Formula C18H37NaO3S
Molecular Weight (g/mol) 356.541
MDL Number MFCD00051526
InChI Key KBAFDSIZQYCDPK-UHFFFAOYSA-M
Synonym sodium 1-octadecanesulfonate, sodium octadecane-1-sulfonate, sodium stearylsulfonate, sodium octadecanesulfonate, 1-octadecanesulfonic acid sodium salt, sodium octadecane-1-sulphonate, 1-octadecanesulfonic acid, sodium salt, sodium sterylsulfonate, sodium oxadecanesulfonate, sodium n-octadecylsulfonate
PubChem CID 23668003
IUPAC Name sodium;octadecane-1-sulfonate
SMILES CCCCCCCCCCCCCCCCCCS(=O)(=O)[O-].[Na+]

Specifications

Color White
Quantity 5 g
Formula Weight 356.54
Physical Form Crystalline Powder
Chemical Name or Material Sodium 1-Octadecanesulfonate
EINECSNumber (2)-1640
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.