accessibility menu, dialog, popup

UserName

Viewing profile as user:
HH2OClClNHONNNS

CAS RN 138982-67-9

IUPAC Name: hydrogen 5-{2-[4-(1,2-benzothiazol-3-yl)piperazin-1-yl]ethyl}-6-chloro-2,3-dihydro-1H-indol-2-one hydrate chloride
Synonyms: zeldox ziprasidone hydrochloride monohydrate ziprasidone hydrochloride hydrate
Molecular Weight (g/mol): 467.41
Molecular Formula: C21H24Cl2N4O2S
InChi Key: ZCBZSCBNOOIHFP-UHFFFAOYSA-N
SMILES: [H+].O.[Cl-].ClC1=CC2=C(CC(=O)N2)C=C1CCN1CCN(CC1)C1=NSC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)