Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Ziprasidone hydrochloride monohydrate

Catalog No. AC462432500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC462432500 250 mg
AC462430010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC462432500 Supplier Thermo Scientific Chemicals Supplier No. 462432500

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138982-67-9
Molecular Formula C21H24Cl2N4O2S
Molecular Weight (g/mol) 467.41
InChI Key ZCBZSCBNOOIHFP-UHFFFAOYSA-N
IUPAC Name hydrogen 5-{2-[4-(1,2-benzothiazol-3-yl)piperazin-1-yl]ethyl}-6-chloro-2,3-dihydro-1H-indol-2-one hydrate chloride
SMILES [H+].O.[Cl-].ClC1=CC2=C(CC(=O)N2)C=C1CCN1CCN(CC1)C1=NSC2=CC=CC=C12

Specifications

Quantity 250 mg
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in dimethyl sulfoxide
Formula Weight 467.41
Chemical Name or Material Ziprasidone hydrochloride monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.