accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaHOONNOHOOOOO

CAS RN 139-33-3

IUPAC Name: disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate
Synonyms: disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate
Molecular Weight (g/mol): 336.21
Molecular Formula: C10H14N2Na2O8
InChi Key: ZGTMUACCHSMWAC-UHFFFAOYSA-L
SMILES: [Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
Quantity 
  • (1)
  • (7)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (1)
  • (1)
  • (11)
Grade 
  • (1)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (10)
  • (1)
  • (1)
  • (6)
  • (1)
  • (7)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (11)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)