Learn More
EDTA disodium salt concentrate with zinc complex added (solution B), For 1L standard solution, for complexometry, Solstice
Shop All Solstice Research Chemicals ProductsDescription
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Physical Form | Liquid |
| CAS Max % | 15.84, 1.19, 6.41 |
| Quantity | 1 Ea. |
| UN Number | NONH for all modes of transport |
| Chemical Name or Material | Ethylenediaminetetraacetic acid disodium salt concentrate with zinc complex added (solution B) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.