CAS RN 1390-65-4
Thermo Scientific Chemicals Carmine, pure, certified, alum lake of carminic acid
CAS: 1390-65-4 Molecular Formula: C22H20O13 Molecular Weight (g/mol): 492.39 MDL Number: MFCD00167028 InChI Key: DGQLVPJVXFOQEV-UHFFFAOYNA-N Synonym: C.I. 75470 PubChem CID: 25113282 IUPAC Name: 3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[(2S,3R,4S,5R,6S)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]anthracene-2-carboxylic acid SMILES: CC1=C(C(O)=O)C(O)=CC2=C1C(=O)C1=C(O)C(C3OC(CO)C(O)C(O)C3O)=C(O)C(O)=C1C2=O
Thermo Scientific Chemicals Carmine, pure, high purity biol.stain, alum lake of carminic acid
CAS: 1390-65-4 Molecular Formula: C22H20O13 Molecular Weight (g/mol): 492.39 MDL Number: MFCD00167028 InChI Key: DGQLVPJVXFOQEV-UHFFFAOYNA-N Synonym: C.I. 75470 PubChem CID: 25113282 IUPAC Name: 3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[(2S,3R,4S,5R,6S)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]anthracene-2-carboxylic acid SMILES: CC1=C(C(O)=O)C(O)=CC2=C1C(=O)C1=C(O)C(C3OC(CO)C(O)C(O)C3O)=C(O)C(O)=C1C2=O
Carmine, Sigma-Aldrich
CAS: 1390-65-4
Carmine, MP Biomedicals™
CAS: 1390-65-4 Molecular Formula: C22H20O13 Molecular Weight (g/mol): 492.39 MDL Number: MFCD00167028 InChI Key: DGQLVPJVXFOQEV-UHFFFAOYNA-N Synonym: Carminic acid,Cochineal,Natural Red 4,Nacarat PubChem CID: 25113282 IUPAC Name: 3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-9,10-dihydroanthracene-2-carboxylic acid SMILES: CC1=C(C(O)=O)C(O)=CC2=C1C(=O)C1=C(O)C(C3OC(CO)C(O)C(O)C3O)=C(O)C(O)=C1C2=O
Carmine Reagent, for Boron determination, Ricca Chemical
For Boron determination
Carmine 40.0+%, TCI America™
CAS: 1390-65-4 Molecular Formula: C22H20O13 Molecular Weight (g/mol): 492.39 MDL Number: MFCD00167028 InChI Key: DGQLVPJVXFOQEV-UHFFFAOYNA-N Synonym: carmine,b rose liquid,cochineal extract lake,carmine coccus cacti l.,cochineal coccus cacti l.,fema no. 2242,fema no. 2330,ccris 1204,alum lake PubChem CID: 25113282 IUPAC Name: 3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-9,10-dihydroanthracene-2-carboxylic acid SMILES: CC1=C(C(O)=O)C(O)=CC2=C1C(=O)C1=C(O)C(C3OC(CO)C(O)C(O)C3O)=C(O)C(O)=C1C2=O
Fisher Science Education™ Carmine
CAS: 1390-65-4 Molecular Formula: C22H20O13 Molecular Weight (g/mol): 492.39 MDL Number: MFCD00167028 InChI Key: DGQLVPJVXFOQEV-UHFFFAOYNA-N Synonym: Aluminum Lake PubChem CID: 25113282 IUPAC Name: 3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-9,10-dihydroanthracene-2-carboxylic acid SMILES: CC1=C(C(O)=O)C(O)=CC2=C1C(=O)C1=C(O)C(C3OC(CO)C(O)C(O)C3O)=C(O)C(O)=C1C2=O
Carmine Powder, Lab Grade, Innovating Science™
CAS: 1390-65-4 Molecular Weight (g/mol): 492.39 g/mol
Carmine (Certified Biological Stain), Fisher Chemical
CAS: 1390-65-4 Molecular Formula: C22H20O13 Molecular Weight (g/mol): 492.39 MDL Number: MFCD00167028 InChI Key: DGQLVPJVXFOQEV-UHFFFAOYNA-N Synonym: Alum Lake,Carminic Acid PubChem CID: 25113282 IUPAC Name: 3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-9,10-dihydroanthracene-2-carboxylic acid SMILES: CC1=C(C(O)=O)C(O)=CC2=C1C(=O)C1=C(O)C(C3OC(CO)C(O)C(O)C3O)=C(O)C(O)=C1C2=O
Carmine, calcium/aluminum lake, Thermo Scientific™
CAS: 1390-65-4 Molecular Formula: C22H20O13 Molecular Weight (g/mol): 492.39 MDL Number: MFCD00167028 InChI Key: DGQLVPJVXFOQEV-UHFFFAOYNA-N Synonym: carmine,b rose liquid,cochineal extract lake,carmine coccus cacti l.,cochineal coccus cacti l.,fema no. 2242,fema no. 2330,ccris 1204,alum lake PubChem CID: 25113282 IUPAC Name: 3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-9,10-dihydroanthracene-2-carboxylic acid SMILES: CC1=C(C(O)=O)C(O)=CC2=C1C(=O)C1=C(O)C(C3OC(CO)C(O)C(O)C3O)=C(O)C(O)=C1C2=O