Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carmine, Sigma-Aldrich

Catalog No. 01259051 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-259-051 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-259-051 Supplier MilliporeSigma Supelco Supplier No. C102225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Carmine has been used for the staining of mammary glands. It has been used for the staining of the chromosomes.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1390-65-4

Specifications

Color Red
Quantity 25 g
Percent Purity ≥42%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.