accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 139015-33-1

IUPAC Name: N-methylpyrrolidin-3-amine
Synonyms: N-methylpyrrolidin-3-amine
Molecular Weight (g/mol): 100.17
Molecular Formula: C5H12N2
InChi Key: NGZYRKGJWYJGRS-UHFFFAOYNA-N
SMILES: CNC1CCNC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results