Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3R)-(+)-3-(Methylamino)pyrrolidine 98.0+%, TCI America™
SDP

Catalog No. M11075G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11075G 5 g
M11071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M11075G Supplier TCI America Supplier No. M11075G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 139015-33-1
Molecular Formula C5H12N2
Molecular Weight (g/mol) 100.165
MDL Number MFCD00191461
InChI Key NGZYRKGJWYJGRS-RXMQYKEDSA-N
PubChem CID 10261056
IUPAC Name (3R)-N-methylpyrrolidin-3-amine
SMILES CNC1CCNC1

Specifications

Color Yellow
Boiling Point 164°C
Quantity 5 g
UN Number 2734
Formula Weight 100.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (3R)-(+)-3-(Methylamino)pyrrolidine
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.