CAS RN 1391-12-4
IUPAC Name:
2-[(6-{[14-({3,4-dihydroxy-6-methyl-5-[(3,4,5-trihydroxy-6-methyloxan-2-yl)oxy]oxan-2-yl}oxy)-6-hydroxy-7,9,13-trimethyl-6-[3-({[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}methyl)but-3-en-1-yl]-5-oxapentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosan-16-yl]oxy}-4,5-dihydroxy-2-methyloxan-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms:
2-[(6-{[14-({3,4-dihydroxy-6-methyl-5-[(3,4,5-trihydroxy-6-methyloxan-2-yl)oxy]oxan-2-yl}oxy)-6-hydroxy-7,9,13-trimethyl-6-[3-({[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}methyl)but-3-en-1-yl]-5-oxapentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosan-16-yl]oxy}-4,5-dihydroxy-2-methyloxan-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol):
1211.35
Molecular Formula:
C57H94O27
InChi Key:
PSJVJZYSECBFHJ-UHFFFAOYNA-N
SMILES:
CC1C2C(CC3C4CCC5CC(CC(OC6OC(C)C(OC7OC(C)C(O)C(O)C7O)C(O)C6O)C5(C)C4CCC23C)OC2OC(C)C(OC3OC(CO)C(O)C(O)C3O)C(O)C2O)OC1(O)CCC(=C)COC1OC(CO)C(O)C(O)C1O
1
–
1
of
1
results
1
MP Biomedicals, Inc Convallamarin, MP Biomedicals
CAS: 1391-12-4 Molecular Formula: C57H94O27 Molecular Weight (g/mol): 1211.352 InChI Key: PSJVJZYSECBFHJ-ZWQMYNLHSA-N PubChem CID: 44135483 SMILES: CC1C2C(CC3C2(CCC4C3CCC5C4(C(CC(C5)OC6C(C(C(C(O6)C)OC7C(C(C(C(O7)CO)O)O)O)O)O)OC8C(C(C(C(O8)C)OC9C(C(C(C(O9)C)O)O)O)O)O)C)C)OC1(CCC(=C)COC1C(C(C(C(O1)CO)O)O)O)O