Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Convallamarin, MP Biomedicals

Catalog No. ICN202920 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN202920 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN202920 Supplier MP Biomedicals, Inc Supplier No. 0520292080

Chemical Identifiers

CAS 1391-12-4
Molecular Formula C57H94O27
Molecular Weight (g/mol) 1211.352
InChI Key PSJVJZYSECBFHJ-ZWQMYNLHSA-N
PubChem CID 44135483
SMILES CC1C2C(CC3C2(CCC4C3CCC5C4(C(CC(C5)OC6C(C(C(C(O6)C)OC7C(C(C(C(O7)CO)O)O)O)O)O)OC8C(C(C(C(O8)C)OC9C(C(C(C(O9)C)O)O)O)O)O)C)C)OC1(CCC(=C)COC1C(C(C(C(O1)CO)O)O)O)O

Specifications

Quantity 100 mg
Formula Weight 430.61
Chemical Name or Material Convallamarin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.