accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 139122-19-3

IUPAC Name: 4-(2-hydroxyethyl)-2,3-dihydro-1H-indol-2-one
Synonyms: 4-(2-hydroxyethyl)-1,3-dihydroindol-2-one
Molecular Weight (g/mol): 177.2
Molecular Formula: C10H11NO2
InChi Key: QRTHVOUKWCEPKJ-UHFFFAOYSA-N
SMILES: OCCC1=C2CC(=O)NC2=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results