Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Hydroxyethyl)oxindole 98.0+%, TCI America™
SDP

Catalog No. H1497200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1497200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H1497200MG Supplier TCI America Supplier No. H1497200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 139122-19-3
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.203
MDL Number MFCD07782133
InChI Key QRTHVOUKWCEPKJ-UHFFFAOYSA-N
Synonym 4-(2-Hydroxyethyl)-2-indolinone
PubChem CID 10943004
IUPAC Name 4-(2-hydroxyethyl)-1,3-dihydroindol-2-one
SMILES C1C2=C(C=CC=C2NC1=O)CCO

Specifications

Melting Point 153°C
Color White-Yellow
Quantity 200 mg
Formula Weight 177.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(2-Hydroxyethyl)oxindole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.