CAS RN 139141-12-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ NF-kB Activation Inhibitor IV
The NF-kB Activation Inhibitor IV, also referenced under CAS 139141-12-1, controls the biological activity of NF-kB. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.