Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ NF-kB Activation Inhibitor IV

Catalog No. 48141210MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-141-210MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 48-141-210MG Supplier MilliporeSigma™ Supplier No. 48141210MG
Add to Cart
Add to Cart

The NF-kB Activation Inhibitor IV, also referenced under CAS 139141-12-1, controls the biological activity of NF-kB. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Description 98% by HPLC
Target Proteins NF-κB Activation
CAS 139141-12-1
Molecular Formula C15H13FO
Formula Weight 228.3g/mol
Solubility DMSO
Content And Storage Protect from light
Form Crystalline Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.