accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO(S)(R)HOOO(R)CH3(R)OH(R)CH3(R)O(S)O(S)O(R)CH3(S)O(S)(R)H3COH(S)OH(S)CH3O(R)(R)HONH3CCH3(E)(E)

CAS RN 1392-21-8

IUPAC Name: 2-[(4R,5S,6S,7R,9R,10R,11E,13E,16R)-6-{[(2S,3R,4R,5S,6R)-5-{[(2S,4R,5S,6S)-4,5-dihydroxy-4,6-dimethyloxan-2-yl]oxy}-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy}-4,10-dihydroxy-5-methoxy-9,16-dimethyl-2-oxo-1-oxacyclohexadeca-11,13-dien-7-yl]acetaldehyde
Synonyms: kitasamycin leucomycin V 吉他霉素
Molecular Weight (g/mol): 701.85
Molecular Formula: C35H59NO13
InChi Key: XYJOGTQLTFNMQG-KJHBSLKPSA-N
SMILES: CO[C@H]1[C@H](O)CC(=O)O[C@H](C)C\C=C\C=C\[C@H](O)[C@H](C)C[C@H](CC=O)[C@@H]1O[C@@H]1O[C@H](C)[C@@H](O[C@H]2C[C@@](C)(O)[C@@H](O)[C@H](C)O2)[C@@H]([C@H]1O)N(C)C
Color 
  • (1)
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results