Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leucomycins, TCI America™
SDP

Catalog No. K00465G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K00465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K00465G Supplier TCI America Supplier No. K00465G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Macrolide antibiotic mixture isolated from Streptomyces kitasatoensis, active against Gram-positive bacteria, spirochetes, and rickettsiae.

Chemical Identifiers

CAS 1392-21-8
Molecular Formula C35H59NO13
Molecular Weight (g/mol) 701.851
MDL Number MFCD00147435
InChI Key XYJOGTQLTFNMQG-XMWOZFRUSA-N
Synonym Kitasamycin
PubChem CID 132274084
IUPAC Name 2-[(4R,6S,9R,10R,11E,13E,16R)-6-[(2S,3R,4R,5S,6R)-5-[(2S,4R,5S,6S)-4,5-dihydroxy-4,6-dimethyloxan-2-yl]oxy-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-4,10-dihydroxy-5-methoxy-9,16-dimethyl-2-oxo-1-oxacyclohexadeca-11,13-dien-7-yl]acetaldehyde
SMILES CC1CC=CC=CC(C(CC(C(C(C(CC(=O)O1)O)OC)OC2C(C(C(C(O2)C)OC3CC(C(C(O3)C)O)(C)O)N(C)C)O)CC=O)C)O

Specifications

Type Leucomycin
Physical Form Crystalline Powder
Color White/Yellow
Quantity 5 g
Percent Purity 48.0% (HPLC)
Grade Not Available
Chemical Name or Material Leucomycins
RTECSNumber OH4550000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.