accessibility menu, dialog, popup

UserName

Viewing profile as user:
FBrBrF

CAS RN 139215-43-3

IUPAC Name: 1,2-dibromo-3,5-difluorobenzene
Synonyms: 1,2-dibromo-3,5-difluorobenzene
Molecular Weight (g/mol): 271.89
Molecular Formula: C6H2Br2F2
InChi Key: GABNJPUNFZFOJE-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C(Br)C(Br)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results