Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dibromo-3,5-difluorobenzene 98.0+%, TCI America™
SDP

Catalog No. D202325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D202325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D202325G Supplier TCI America Supplier No. D202325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 139215-43-3
Molecular Formula C6H2Br2F2
Molecular Weight (g/mol) 271.887
MDL Number MFCD00042559
InChI Key GABNJPUNFZFOJE-UHFFFAOYSA-N
Synonym benzene, 1,2-dibromo-3,5-difluoro, 1,2-dibromo-3,5-difluoro-benzene, pubchem3438, acmc-1c3vh, 2,3-dibromo-1,5-difluorobenzene, 1,2-bis bromanyl-3,5-bis fluoranyl benzene
PubChem CID 2724517
IUPAC Name 1,2-dibromo-3,5-difluorobenzene
SMILES C1=C(C=C(C(=C1F)Br)Br)F

Specifications

Melting Point 37°C
Quantity 25 g
Formula Weight 271.89
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Dibromo-3,5-difluorobenzene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.