accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3IIH3C

CAS RN 1392203-19-8

IUPAC Name: 5,5'-diiodo-2,2'-dimethyl-1,1'-biphenyl
Synonyms: 5,5'-diiodo-2,2'-dimethyl-1,1'-biphenyl
Molecular Weight (g/mol): 434.06
Molecular Formula: C14H12I2
InChi Key: VBRWVADZBWZCMH-UHFFFAOYSA-N
SMILES: CC1=C(C=C(I)C=C1)C1=C(C)C=CC(I)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results