accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OONONCH3OCH3

CAS RN 139290-70-3

IUPAC Name: tert-butyl 4-[methoxy(methyl)carbamoyl]piperidine-1-carboxylate
Synonyms: tert-butyl 4-[methoxy(methyl)carbamoyl]piperidine-1-carboxylate
Molecular Weight (g/mol): 272.35
Molecular Formula: C13H24N2O4
InChi Key: ITCQNWXLNZGEHP-UHFFFAOYSA-N
SMILES: CON(C)C(=O)C1CCN(CC1)C(=O)OC(C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results