Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl 4-(N-Methoxy-N-methylcarbamoyl)-1-piperidinecarboxylate 98.0+%, TCI America™
SDP

Catalog No. B37081G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B37081G Supplier TCI America Supplier No. B37081G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 139290-70-3
Molecular Formula C13H24N2O4
Molecular Weight (g/mol) 272.35
MDL Number MFCD07368262
InChI Key ITCQNWXLNZGEHP-UHFFFAOYSA-N
Synonym 4-(N-Methoxy-N-methylcarbamoyl)-1-piperidinecarboxylic Acid tert-Butyl Ester, 1-(tert-Butoxycarbonyl)-N-methoxy-N-methyl-4-piperidinecarboxamide, 1-Boc-N-methoxy-N-methyl-4-piperidinecarboxamide, 1-(tert-Butoxycarbonyl)-4-(N-methoxy-N-methylcarbamoyl)pipe
PubChem CID 11065582
IUPAC Name tert-butyl 4-[methoxy(methyl)carbamoyl]piperidine-1-carboxylate
SMILES CON(C)C(=O)C1CCN(CC1)C(=O)OC(C)(C)C

Specifications

Melting Point 73°C
Color White
Boiling Point 150°C
Quantity 1 g
Formula Weight 272.35
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material tert-Butyl 4-(N-Methoxy-N-methylcarbamoyl)-1-piperidinecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.