accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13934-03-7

IUPAC Name: (2S)-2-({[(4E)-5-(hydroxymethyl)-2-methyl-3-oxo-3,4-dihydropyridin-4-ylidene]methyl}amino)pentanedioic acid
Synonyms: (2S)-2-({[(4E)-5-(hydroxymethyl)-2-methyl-3-oxopyridin-4-ylidene]methyl}amino)pentanedioic acid
Molecular Weight (g/mol): 296.28
Molecular Formula: C13H16N2O6
InChi Key: XKBXXIDGNQTCSE-CYNRKNSPSA-N
SMILES: CC1=NC=C(CO)\C(=C/N[C@@H](CCC(O)=O)C(O)=O)C1=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results