Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridoxylidene-L-glutamic Acid Dipotassium Salt, TCI America™
SDP

Catalog No. P09881G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P09881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P09881G Supplier TCI America Supplier No. P09881G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13934-03-7
Molecular Formula C13H14K2N2O6
Molecular Weight (g/mol) 372.46
MDL Number MFCD06809726
InChI Key JCHMAGPATAIHHG-IJCFLWNESA-L
PubChem CID 121233270
IUPAC Name dipotassium;(2S)-2-[[(E)-[5-(hydroxymethyl)-2-methyl-3-oxopyridin-4-ylidene]methyl]amino]pentanedioate
SMILES CC1=NC=C(C(=CNC(CCC(=O)[O-])C(=O)[O-])C1=O)CO.[K+].[K+]

Specifications

Quantity 1 g
Formula Weight 372.46
Physical Form Crystalline Powder
Chemical Name or Material Pyridoxylidene-L-glutamic Acid Dipotassium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.