accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 139404-48-1

IUPAC Name: (1R,2S,4R,5S)-7-{[2-hydroxy-2,2-bis(thiophen-2-yl)acetyl]oxy}-9,9-dimethyl-3-oxa-9-azatricyclo[3.3.1.0²,⁴]nonan-9-ium bromide
Synonyms: tiotropium bromide
Molecular Weight (g/mol): 472.41
Molecular Formula: C19H22BrNO4S2
InChi Key: DQHNAVOVODVIMG-CJTHRXGHSA-M
SMILES: [Br-].C[N+]1(C)[C@H]2CC(C[C@@H]1[C@@H]1O[C@H]21)OC(=O)C(O)(C1=CC=CS1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)