CAS RN 13943-58-3
IUPAC Name:
tetrapotassium ironhexacarbonitrile
Synonyms:
potassium ferrocyanide
Molecular Weight (g/mol):
374.39
Molecular Formula:
C6H6FeK4N6
InChi Key:
FIBBHPJORRWWPX-UHFFFAOYSA-N
SMILES:
[K+].[K+].[K+].[K+].[Fe].C#N.C#N.C#N.C#N.C#N.C#N
1
–
1
of
1
results
1
Potassium hexacyanoferrate(II), 0.1N Standardized Solution
CAS: 13943-58-3 MDL Number: MFCD00167022 Synonym: Potassium ferrocyanide