Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium hexacyanoferrate(II), 0.1N Standardized Solution

Catalog No. AA35585K2
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA35585K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA35585K2 Supplier Thermo Scientific Chemicals Supplier No. 035585K2
Add to Cart

CAS: 13943-58-3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13943-58-3
MDL Number MFCD00167022
Synonym Potassium ferrocyanide

Specifications

Color Yellow
Linear Formula K4Fe(CN)6
Concentration 0.1N
Merck Index 14,7631
Formula Weight 368.36
Odor Odorless
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Potassium hexacyanoferrate(II), Standardized Solution
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.