accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 139680-47-0

IUPAC Name: N,N-bis(2-methylpropyl)-4-nitrobenzamide
Synonyms: N,N-bis(2-methylpropyl)-4-nitrobenzamide
Molecular Weight (g/mol): 278.35
Molecular Formula: C15H22N2O3
InChi Key: IFYJJQOERYQZBL-UHFFFAOYSA-N
SMILES: CC(C)CN(CC(C)C)C(=O)C1=CC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results