accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13980-76-2

IUPAC Name: 1-(4-methoxyphenyl)-2,5-dihydro-1H-1,2,3,4-tetrazole-5-thione
Synonyms: 1-(4-methoxyphenyl)-2H-1,2,3,4-tetrazole-5-thione
Molecular Weight (g/mol): 208.24
Molecular Formula: C8H8N4OS
InChi Key: UFALKIBIWOKBDL-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)N1NN=NC1=S
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results