Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Mercapto-1-(4-methoxyphenyl)-1H-tetrazole 98.0+%, TCI America™
SDP

Catalog No. M129210G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M129210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M129210G Supplier TCI America Supplier No. M129210G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13980-76-2
Molecular Formula C8H8N4OS
Molecular Weight (g/mol) 208.239
MDL Number MFCD01321180
InChI Key UFALKIBIWOKBDL-UHFFFAOYSA-N
Synonym 1-(4-Methoxyphenyl)-1H-tetrazole-5-thiol
PubChem CID 695766
IUPAC Name 1-(4-methoxyphenyl)-2H-tetrazole-5-thione
SMILES COC1=CC=C(C=C1)N2C(=S)N=NN2

Specifications

Color White-Yellow
Quantity 10 g
UN Number 1325
Formula Weight 208.24
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Mercapto-1-(4-methoxyphenyl)-1H-tetrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.