accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3N

CAS RN 13989-82-7

IUPAC Name: 4-(dimethylamino)butanenitrile
Synonyms: 4-(dimethylamino)butanenitrile
Molecular Weight (g/mol): 112.18
Molecular Formula: C6H12N2
InChi Key: HCLFLZTVKYHLCF-UHFFFAOYSA-N
SMILES: CN(C)CCCC#N
Grade 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results