Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Dimethylaminobutyronitrile 98.0+%, TCI America™
SDP

Catalog No. D45461ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45461ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D45461ML Supplier TCI America Supplier No. D45461ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Organic chemical building block for laboratory research.

Chemical Identifiers

CAS 13989-82-7
Molecular Formula C6H12N2
Molecular Weight (g/mol) 112.176
MDL Number MFCD00019892
InChI Key HCLFLZTVKYHLCF-UHFFFAOYSA-N
PubChem CID 56430
IUPAC Name 4-(dimethylamino)butanenitrile
SMILES CN(C)CCCC#N

Specifications

Color Yellow
Boiling Point 80°C
Quantity 1 mL
UN Number 1993
Formula Weight 112.18
Percent Purity ≥98.0% (GC,T)
Grade Not Available
Physical Form Liquid
Chemical Name or Material 4-Dimethylaminobutyronitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.