accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaNaNaNaOONNOOOONOOOO

CAS RN 140-01-2

IUPAC Name: pentasodium 2-[bis({2-[bis(carboxylatomethyl)amino]ethyl})amino]acetate
Synonyms: pentasodium pentetate diethylenetriaminepentaacetic acid, penta sodiu*
Molecular Weight (g/mol): 503.26
Molecular Formula: C14H18N3Na5O10
InChi Key: LQPLDXQVILYOOL-UHFFFAOYSA-I
SMILES: [Na+].[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CCN(CC([O-])=O)CC([O-])=O
Molecular Weight (g/mol) 
  • (9)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (3)
  • (3)
Percent Purity 
  • (6)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (9)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (6)
  • (2)
  • (3)
  • (3)