Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylenetriaminepentaacetic Acid, Na, 40% Solution, MP Biomedicals

Catalog No. ICN206940 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN206940 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN206940 Supplier MP Biomedicals, Inc Supplier No. 0520694080

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 140-01-2
Molecular Formula C14H18N3Na5O10
Molecular Weight (g/mol) 503.26
MDL Number MFCD00051016
InChI Key LQPLDXQVILYOOL-UHFFFAOYSA-I
Synonym pentasodium dtpa, tetralon b, trilon c, versenex 80, pentasodium pentetate, hamp-ex 80, detarex py, kiresuto p, chelest p, plexene d
PubChem CID 8779
IUPAC Name pentasodium 2-[bis({2-[bis(carboxylatomethyl)amino]ethyl})amino]acetate
SMILES [Na+].[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CCN(CC([O-])=O)CC([O-])=O

Specifications

Formula Weight 503.26
Quantity 100 g
Chemical Name or Material Diethylenetriamine Pentaacetic Acid, Na, 40% Soln.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.