accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3OCH3O(Z)

CAS RN 140-03-4

IUPAC Name: methyl (9Z)-12-(acetyloxy)octadec-9-enoate
Synonyms: methyl acetyl ricinoleate methylester kyseliny acetylricinolejove acetate methyl ricinoleate flexricin P-4 ricinoleic acid, methyl ester, acetate methyl ricinoleate, acetate
Molecular Weight (g/mol): 354.53
Molecular Formula: C21H38O4
InChi Key: CMOYPQWMTBSLJK-KAMYIIQDNA-N
SMILES: CCCCCCC(C\C=C/CCCCCCCC(=O)OC)OC(C)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)