Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl O-Acetylricinoleate 80.0+%, TCI America™
SDP

Catalog No. A0879500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0879500ML 500 mL
A087925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0879500ML Supplier TCI America Supplier No. A0879500ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 140-03-4
Molecular Formula C21H38O4
Molecular Weight (g/mol) 354.53
MDL Number MFCD00026204
InChI Key CMOYPQWMTBSLJK-KAMYIIQDNA-N
Synonym O-Acetylricinoleic Acid Methyl Ester, Methyl (Z)-12-Acetoxy-9-octadecenoate
PubChem CID 27396493
IUPAC Name methyl (9Z)-12-(acetyloxy)octadec-9-enoate
SMILES CCCCCCC(C\C=C/CCCCCCCC(=O)OC)OC(C)=O

Specifications

Color Yellow
Boiling Point 260°C
Quantity 500 mL
Formula Weight 354.53
Percent Purity ≥80.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl O-Acetylricinoleate
EINECSNumber (2)-1339
RTECSNumber VJ3410000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.