accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO

CAS RN 140-25-0

IUPAC Name: benzyl dodecanoate
Synonyms: benzyl laurate
Molecular Weight (g/mol): 290.45
Molecular Formula: C19H30O2
InChi Key: QNRYOQRUGRVBRL-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results