Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl Laurate 98.0+%, TCI America™
SDP

Catalog No. L010725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L010725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L010725ML Supplier TCI America Supplier No. L010725ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 140-25-0
Molecular Formula C19H30O2
Molecular Weight (g/mol) 290.447
MDL Number MFCD00026558
InChI Key QNRYOQRUGRVBRL-UHFFFAOYSA-N
Synonym Lauric Acid Benzyl Ester
PubChem CID 8791
IUPAC Name benzyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 10°C
Color Yellow
Boiling Point 211°C
Quantity 25 mL
Formula Weight 290.45
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl Laurate
RTECSNumber JR3425000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.