accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 140-28-3

IUPAC Name: benzyl[2-(benzylamino)ethyl]amine
Synonyms: benzathine DBED
Molecular Weight (g/mol): 240.35
Molecular Formula: C16H20N2
InChi Key: JUHORIMYRDESRB-UHFFFAOYSA-N
SMILES: C(CNCC1=CC=CC=C1)NCC1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)