accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14001-66-2

IUPAC Name: 5-bromo-2-methoxypyrimidine
Synonyms: 5-bromo-2-methoxypyrimidine
Molecular Weight (g/mol): 189.01
Molecular Formula: C5H5BrN2O
InChi Key: DWVCZDMMGYIULX-UHFFFAOYSA-N
SMILES: COC1=NC=C(Br)C=N1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)