accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14017-41-5

IUPAC Name: λ²-cobalt(2+) disulfamate
Synonyms: λ²-cobalt(2+) disulfamate
Molecular Weight (g/mol): 251.09
Molecular Formula: CoH4N2O6S2
InChi Key: WLQXLCXXAPYDIU-UHFFFAOYSA-L
SMILES: [Co++].NS([O-])(=O)=O.NS([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results