accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOONOOONOOOY

CAS RN 140191-40-8

IUPAC Name: yttrium(3+) trinitrate
Synonyms: yttrium(III) nitrate
Molecular Weight (g/mol): 274.92
Molecular Formula: N3O9Y
InChi Key: BXJPTTGFESFXJU-UHFFFAOYSA-N
SMILES: [Y+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Grade 
  • (4)
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)