accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14024-61-4

IUPAC Name: palladium(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Synonyms: palladium(II) acetylacetonate
Molecular Weight (g/mol): 304.64
Molecular Formula: C10H14O4Pd
InChi Key: JKDRQYIYVJVOPF-FDGPNNRMSA-L
SMILES: [Pd++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (3)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results