accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14027-75-9

IUPAC Name: 3-fluoro-5-nitrobenzoic acid
Synonyms: 3-fluoro-5-nitrobenzoic acid
Molecular Weight (g/mol): 185.11
Molecular Formula: C7H4FNO4
InChi Key: VIPUIECMSDQUIK-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=CC(F)=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results