Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluoro-5-nitrobenzoic acid, 97%, Thermo Scientific™

Catalog No. AAH6329503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6329503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6329503 Supplier Thermo Scientific Chemicals Supplier No. H6329503

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14027-75-9
Molecular Formula C7H4FNO4
Molecular Weight (g/mol) 185.11
MDL Number MFCD05864360
InChI Key VIPUIECMSDQUIK-UHFFFAOYSA-N
Synonym 3-fluoro-5-nitrobenzoic acid, benzoic acid, 3-fluoro-5-nitro, 5-fluoro-3-nitrobenzoic acid, pubchem1400, intermediates-zcf02073, spec-inf-bb s2256
PubChem CID 2759047
SMILES C1=C(C=C(C=C1[N+](=O)[O-])F)C(=O)O

Specifications

Quantity 1 g
Formula Weight 185.11
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-Fluoro-5-nitrobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.