accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOBOHOHO

CAS RN 14047-29-1

IUPAC Name: 4-(dihydroxyboranyl)benzoic acid
Synonyms: 4-carboxyphenylboronic acid
Molecular Weight (g/mol): 165.94
Molecular Formula: C7H7BO4
InChi Key: SIAVMDKGVRXFAX-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=C1)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (7)
  • (2)
Quantity 
  • (4)
  • (1)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (6)
  • (1)
  • (7)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)