Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Carboxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. C13535G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1353-5G 5 g
C13531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1353-5G Supplier TCI America Supplier No. C13535G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14047-29-1
Molecular Formula C7H7BO4
MDL Number MFCD00151801
InChI Key SIAVMDKGVRXFAX-UHFFFAOYSA-N
PubChem CID 312183

Specifications

Color White-Yellow
Quantity 5 g
Synonym 4-carboxyphenylboronic acid, 4-carboxybenzeneboronic acid, 4-dihydroxyboranyl benzoic acid, benzoic acid, 4-borono, 4-dihydroxyboryl benzoic acid
SMILES B(C1=CC=C(C=C1)C(=O)O)(O)O
IUPAC Name 4-boronobenzoic acid
Molecular Weight (g/mol) 165.939
Formula Weight 165.94
Physical Form Crystalline Powder
RTECSNumber CY8925000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.