accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaH3COO

CAS RN 14047-60-0

IUPAC Name: sodium nonanoate
Synonyms: sodium nonoate
Molecular Weight (g/mol): 180.22
Molecular Formula: C9H17NaO2
InChi Key: LTOCMXUTASYUOC-UHFFFAOYSA-M
SMILES: [Na+].CCCCCCCCC([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results