Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Nonanoate 98.0+%, TCI America™
SDP

Catalog No. N029125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N029125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N029125G Supplier TCI America Supplier No. N029125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14047-60-0
Molecular Formula C9H17NaO2
Molecular Weight (g/mol) 180.223
MDL Number MFCD00069612
InChI Key LTOCMXUTASYUOC-UHFFFAOYSA-M
Synonym Pelargonic Acid Sodium Salt, Nonanoic Acid Sodium Salt, Sodium Pelargonate
PubChem CID 23674758
IUPAC Name sodium;nonanoate
SMILES CCCCCCCCC(=O)[O-].[Na+]

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 180.22
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Nonanoate
EINECSNumber (2)-0611&(7)-0973
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.