accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COCH3HOCH2

CAS RN 14049-11-7

IUPAC Name: 6-ethenyl-2,2,6-trimethyloxan-3-ol
Synonyms: 6-ethenyl-2,2,6-trimethyloxan-3-ol
Molecular Weight (g/mol): 170.25
Molecular Formula: C10H18O2
InChi Key: BCTBAGTXFYWYMW-UHFFFAOYNA-N
SMILES: CC1(C)OC(C)(CCC1O)C=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results